Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl-beta-D-galactopyranoside hydrate, 98+%

Catalog No. p-7046041
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1291103 1 g
AAL1291106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1291103 Supplier Thermo Scientific Chemicals Supplier No. L1291103
Only null left
Add to Cart

CAS: 200422-18-0 | C12H15NO8 | 301.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 200422-18-0
Molecular Formula C12H15NO8
Molecular Weight (g/mol) 301.25
MDL Number MFCD00063256
InChI Key IFBHRQDFSNCLOZ-MFZRPPIDNA-N
Synonym 4-nitrophenyl-beta-d-galactopyranoside hydrate, 2r,3r,4s,5r,6s-2-hydroxymethyl-6-4-nitrophenoxy tetrahydro-2h-pyran-3,4,5-triol hydrate, p-nitrophenyl-?-d-galactoside hydrate, 4-nitrophenyl beta-d-galactopyranoside hydrate
PubChem CID 16218614
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Melting Point 180°C to 184°C
Quantity 1 g
Beilstein 92213
Optical Rotation −84° (c=0.3 in Water)
Formula Weight 319.27 (301.25 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 4-Nitrophenyl-beta-D-galactopyranoside hydrate
EINECSNumber 221-584-5
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.