Learn More
4-Nitrophenyl-beta-D-glucopyranoside, 98+%
Description
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrophenyl-β-D-glucopyranoside acts as a chromogenic beta-D-glucosidase substrate used for the measurement of beta-glucosidase activity. Further, it is used in research, biochemical enzyme assays and in vitro diagnostic analysis.
Solubility
Soluble in water, methanol, warm ethanol, dimethyl sulfoxide and dimethylformamide. Insoluble in ether.
Notes
Store in a cool place. Incompatible with strong bases.
Specifications
Specifications
| Melting Point | 166°C to 167°C |
| Quantity | 1 g |
| Beilstein | 92211 |
| Solubility Information | Soluble in water,methanol,warm ethanol,dimethyl sulfoxide and dimethylformamide. Insoluble in ether. |
| Optical Rotation | −103° (c=1 in Water) |
| Formula Weight | 301.25 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Nitrophenyl-beta-D-glucopyranoside |
Safety and Handling
| EINECSNumber | 219-661-3 |
| TSCA | Yes |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.