Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl-beta-D-glucuronide, 99+%

Catalog No. AC336790010
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 10344-94-2 | C12H12NO9 | 314.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10344-94-2
Molecular Formula C12H12NO9
Molecular Weight (g/mol) 314.23
MDL Number MFCD00036210
InChI Key QSUILVWOWLUOEU-GOVZDWNOSA-M
Synonym 4-nitrophenyl-beta-d-glucuronide, 4-nitrophenylglucuronide, 4-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl beta-d-glucuronide, p-nitrophenyl beta-d-glucopyranosiduronic acid, 4-nitrophenyl b-d-glucosiduronic acid, p-nitrophenyl-b-d-glucuronide, p-nitrophenyl beta-d-glucuronide, beta-d-glucopyranosiduronic acid, 4-nitrophenyl, labeled with carbon-14, 4-nitrophenyl-beta-d-glucopyranosiduronic acid
PubChem CID 82565
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 0.05% max. (p-Nitrophenol) (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation −107° to −113° (20°C, 589nm) (c=1, H2O)
Solubility Information (1% in water) Clear colorless to light yellow
Formula Weight 315.24
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 4-Nitrophenyl-β-D-glucuronide
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 233-753-0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.