Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Nitrophenyl-beta-D-glucuronide, 99+%
Click to view available options
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Color | Colorless to Yellow |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 0.05% max. (p-Nitrophenol) (HPLC) |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Specific Rotation | −107° to −113° (20°C, 589nm) (c=1, H2O) |
| Solubility Information | (1% in water) Clear colorless to light yellow |
| Formula Weight | 315.24 |
| Show More |
Safety and Handling
| Hazard Category | MOISTURE SENSITIVE |
| EINECSNumber | 233-753-0 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction