Learn More
4-Nitrophenyl chloroformate, 97%
Description
4-Nitrophenyl chloroformate is used in the synthesis of poly(oxyethylene) modified dextrans. It is also used to prepare a cleavable, heterobifunctional cross-linker for reversible conjugation of amines to thiol-modified DNA. Further, it is used in the synthesis of mixed, activated carbonates, thiocarbonates and activated urethanes. In addition to this, it is used in the preparation of 4-nitrophenyl diazoacetate with diazomethane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Nitrophenyl chloroformate is used in the synthesis of poly(oxyethylene) modified dextrans. It is also used to prepare a cleavable, heterobifunctional cross-linker for reversible conjugation of amines to thiol-modified DNA. Further, it is used in the synthesis of mixed, activated carbonates, thiocarbonates and activated urethanes. In addition to this, it is used in the preparation of 4-nitrophenyl diazoacetate with diazomethane.
Solubility
Soluble in acetone, chloroform, acetone, toluene and benzene.
Notes
Moisture sensitive. Store in a cool place. Incompatible with water, acids, alcohols, ammonia, amines and strong bases.
Specifications
Specifications
| Melting Point | 75°C to 82°C |
| Boiling Point | 159°C to 162°C (19 mmHg) |
| Flash Point | >110°C (230°F) |
| Odor | Acrid |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 518127 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in acetone,chloroform,acetone,toluene and benzene. |
| Formula Weight | 201.57 |
| Show More |
Safety and Handling
| Hazard Category | H301+H311+H331-H314-H335 |
| Hazard Statement | GHS H Statement H301-H314-H318-H332 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S. |
| EINECSNumber | 231-706-9 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.