Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl phosphate disodium salt hexahydrate, 96%

Catalog No. p-7082006
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 333338-18-4 | C6H4NNa2O6P | 263.05 g/mol

Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substrate for alkaline and acid phosphatase determinations4-Nitrophenyl phosphate disodium salt hexahydrate is a chromogenic substrate used in the determination of acid and alkaline phosphatases.

Solubility
Miscible with water. Immiscible with ethanol and acetone.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333338-18-4
Molecular Formula C6H4NNa2O6P
Molecular Weight (g/mol) 263.05
MDL Number MFCD00007319
InChI Key VIYFPAMJCJLZKD-UHFFFAOYSA-L
Synonym pnpp, disodium 4-nitrophenylphosphate, sodium 4-nitrophenyl phosphate, disodium 4-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, disodium salt, disodium p-nitrophenyl phosphate, phosphoric acid, mono 4-nitrophenyl ester, sodium salt 1:2, p-nitrophenyl phosphate, pnpp liquid substrate system
PubChem CID 77949
IUPAC Name disodium;(4-nitrophenyl) phosphate
SMILES [Na+].[Na+].[O-][N+](=O)C1=CC=C(OP([O-])([O-])=O)C=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 5324661
Solubility Information Miscible with water. Immiscible with ethanol and acetone.
Formula Weight 371.15 (263.06 Anhydrous)
Percent Purity 98%
Chemical Name or Material 4-Nitrophenyl phosphate disodium salt hexahydrate
EINECSNumber 224-246-5
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.