Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylacetone, 98%

Catalog No. AAB2118306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2118306 5 g
AAB2118303 1 g
AAB2118314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2118306 Supplier Thermo Scientific Chemicals Supplier No. B2118306
Add to Cart

CAS: 5332-96-7 | C9H9NO3 | 179.175 g/mol

It is applied as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an intermediate in organic synthesis.

Solubility
Insoluble in water, soluble in usual organic solvents.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5332-96-7
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00051518
InChI Key GEWWCWZGHNIUBW-UHFFFAOYSA-N
Synonym 4-nitrophenylacetone, 1-4-nitrophenyl propan-2-one, 2-propanone, 1-4-nitrophenyl, 1-4-nitrophenyl-2-propanone, 1-4-nitrophenyl acetone, p-nitrophenylazeton, p-nitrophenylacetone, pubchem18646, 4-nitrophenyl acetone, acmc-1az6k
PubChem CID 219367
IUPAC Name 1-(4-nitrophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 61°C to 64°C
Quantity 5 g
Beilstein 1953054
Solubility Information Insoluble in water,soluble in usual organic solvents.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Nitrophenylacetone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.