Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenylhydrazine 98%, Thermo Scientific™

Catalog No. AC151521000
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-16-3
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007579
InChI Key KMVPXBDOWDXXEN-UHFFFAOYSA-N
Synonym 4-nitrophenyl hydrazine, p-nitrophenylhydrazine, hydrazine, 4-nitrophenyl, p-hydrazinonitrobenzene, hydrazine, p-nitrophenyl, 1-4-nitrophenyl hydrazine, ccris 3139, p-nitrophenyl hydrazine, pnd, p-nitrophenylhydrazin
PubChem CID 3310889
ChEBI CHEBI:66931
IUPAC Name (4-nitrophenyl)hydrazine
SMILES NNC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Linear Formula O2NC6H4NHNH2
Quantity 100 g
Formula Weight 153.14
Chemical Name or Material 4-Nitrophenylhydrazine, 98%
EINECSNumber 202-824-8
RTECSNumber MV8225000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.