Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrotoluene, 99%

Catalog No. AC129050010
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass bottle

CAS: 99-99-0 | C7H7NO2 | 137.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-99-0
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007366
InChI Key ZPTVNYMJQHSSEA-UHFFFAOYSA-N
Synonym 4-nitrotoluene, p-nitrotoluene, 4-methylnitrobenzene, 4-nitrotoluol, benzene, 1-methyl-4-nitro, toluene, p-nitro, p-methylnitrobenzene, para-nitrotoluol, nitrotoluenos, nitrotoluene, p
PubChem CID 7473
ChEBI CHEBI:35227
IUPAC Name 1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 51.0°C to 54.0°C
Color Yellow
Boiling Point 238.0°C
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4NO2
Quantity 1 kg
Beilstein 05, 323
Merck Index 15, 6737
Solubility Information Solubility in water: 0.35g/L (20°C). Other solubilities: soluble in ethanol, ether, benzene, chloroform and, acetone
Formula Weight 137.14
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Nitrotoluene, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a po
EINECSNumber 202-808-0
RTECSNumber XT3325000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.