Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrotriphenylamine 98.0+%, TCI America™
SDP

Catalog No. n08311g
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 4316-57-8
Molecular Formula C18H14N2O2
Molecular Weight (g/mol) 290.322
MDL Number MFCD00814232
InChI Key UQOKZDUUBVGFAK-UHFFFAOYSA-N
PubChem CID 350141
IUPAC Name 4-nitro-N,N-diphenylaniline
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)C3=CC=C(C=C3)[N+](=O)[O-]

Specifications

Melting Point 145°C
Quantity 1 g
Formula Weight 290.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrotriphenylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.