Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Octyne 99.0+%, TCI America™
SDP

Catalog No. p-7134199
Click to view available options
Quantity:
5 mL
25 mL

Chemical Identifiers

CAS 1942-45-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.2
MDL Number MFCD00009471
InChI Key GZTNBKQTTZSQNS-UHFFFAOYSA-N
Synonym 4-octyne, dipropylacetylene, 1,2-dipropylacetylene, di-n-propylacetylene, acmc-209ey0, 4-octyne 5g, n-c3h7c.$.cc3h7, 3,3?-dihexyloxacarbocyanine iodide
PubChem CID 16029
IUPAC Name oct-4-yne
SMILES CCCC#CCCC

Specifications

Melting Point -103°C
Color Colorless
Boiling Point 131°C
Quantity 5 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Octyne
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.