Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Oxo-1-phenylcyclohexanecarbonitrile 98.0+%, TCI America™
SDP

Catalog No. O047025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O047025G 25 g
O04705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O047025G Supplier TCI America Supplier No. O047025G

Chemical Identifiers

CAS 25115-74-6
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.25
MDL Number MFCD00044816
InChI Key GKXOABVSZWCJJK-UHFFFAOYSA-N
Synonym 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone
PubChem CID 91282
IUPAC Name 4-oxo-1-phenylcyclohexane-1-carbonitrile
SMILES O=C1CCC(CC1)(C#N)C1=CC=CC=C1

Specifications

Melting Point 119°C
Color White
Quantity 25 g
Formula Weight 199.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Oxo-1-phenylcyclohexanecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.