Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-oxo-4-(2-thienyl)butanoic acid, Thermo Scientific™

Catalog No. BTB10644EA
Click to view available options
Quantity:
1 g
10 g
25 g

Chemical Identifiers

CAS 4653-08-1
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.21
InChI Key ULJMYWHLMLRYSO-UHFFFAOYSA-N
Synonym 3-2-thenoyl propionic acid, 4-oxo-4-thiophen-2-yl butanoic acid, 4-oxo-4-2-thienyl butanoic acid, 4-oxo-4-2-thienyl butyric acid, 3-2-thenoyl propanoic acid, 4-thien-2-yl-4-oxobutyric acid, 4-oxo-4-thien-2-yl butyric acid, 4-thien-2-yl-4-oxobutanoic acid, 4-oxo-4-thien-2-yl butanoic acid, 4-oxo-4-thiophen-2-yl-butyric acid
PubChem CID 78385
ChEBI CHEBI:64434
IUPAC Name 4-oxo-4-thiophen-2-ylbutanoic acid
SMILES C1=CSC(=C1)C(=O)CCC(=O)O

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material 4-Oxo-4-(2-thienyl)butanoic acid
Hazard Category GHS Signal Word: Warning
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.