Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Penten-1-ol 98.0+%, TCI America™

Click to view available options
Quantity:
25 mL
250 mL
Specifications
Specifications
Color | Colorless |
Boiling Point | 138°C |
Quantity | 250 mL |
UN Number | 1987 |
Formula Weight | 86.13 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | 4-Penten-1-ol |
Safety and Handling
EINECSNumber | (2)-2393 |
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction