Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyaniline, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
139-59-3
C12H11NO
185.226
MFCD00007862
WOYZXEVUWXQVNV-UHFFFAOYSA-N
p-phenoxyaniline,4-aminodiphenyl ether,benzenamine, 4-phenoxy,4-phenoxybenzenamine,4-aminophenyl phenyl ether,4-aminodifenylether,4-amino-1-phenoxybenzene,aniline, p-phenoxy,4-aminobiphenyl ether,4-aminodiphenylether
8764
4-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=C(C=C2)N
139-59-3
C12H11NO
185.226
MFCD00007862
WOYZXEVUWXQVNV-UHFFFAOYSA-N
p-phenoxyaniline,4-aminodiphenyl ether,benzenamine, 4-phenoxy,4-phenoxybenzenamine,4-aminophenyl phenyl ether,4-aminodifenylether,4-amino-1-phenoxybenzene,aniline, p-phenoxy,4-aminobiphenyl ether,4-aminodiphenylether
8764
4-phenoxyaniline
C1=CC=C(C=C1)OC2=CC=C(C=C2)N
Color Brown to Brown-Green
Quantity 5g
Melting Point 82°C to 86°C
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5OC6H4NH2
Beilstein 13, 438
Formula Weight 185.23
Physical Form Crystalline Flakes or Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Phenoxyaniline
Show More Show Less

GHS H Statement
Harmful if swallowed. May cause an allergic skin reaction. Very toxic to aquatic life with long lasting effects. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water.

Warning