Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P125625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2215-77-2
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
MDL Number MFCD00002539
InChI Key RYAQFHLUEMJOMF-UHFFFAOYSA-N
Synonym p-phenoxybenzoic acid, benzoic acid, 4-phenoxy, diphenyl ether 4-carboxylic acid, 4-phenoxy-benzoic acid, 4-carboxybiphenyl ether, 4-carboxydiphenyl ether, benzoic acid, p-phenoxy, p-carboxydiphenyl ether, 4-phenoxy benzoic acid, 4-phoc6h4cooh
PubChem CID 75182
ChEBI CHEBI:72632
IUPAC Name 4-phenoxybenzoic acid
SMILES OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 214.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Phenoxybenzoic Acid
RTECSNumber DH6293620
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.