Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P197425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Specifications

CAS 51067-38-0
Melting Point 115°C
Molecular Formula C12H11BO3
Quantity 25 g
MDL Number MFCD00093312
Synonym 4-phenoxyphenyl boronic acid, 4-phenoxybenzeneboronic acid, 4-phenoxyphenyl boranediol, boronic acid, 4-phenoxyphenyl, p-phenoxyphenyl boronic acid
InChI Key KFXUHRXGLWUOJT-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)OC2=CC=CC=C2)(O)O
IUPAC Name (4-phenoxyphenyl)boronic acid
Molecular Weight (g/mol) 214.027
PubChem CID 2734377
Formula Weight 214.03
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.