Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1-butanol, 97%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
3360-41-6
C10H14O
150.22
MFCD00002971
LDZLXQFDGRCELX-UHFFFAOYSA-N
4-phenyl-1-butanol,benzenebutanol,4-phenylbutanol,phenylbutyl alcohol,4-phenylbutyl alcohol,1-butanol, 4-phenyl,4-phenyl butanol-1,4-phenyl butanol,unii-q5orz1321g,4-phenyl-n-butanol
76889
4-phenylbutan-1-ol
C1=CC=C(C=C1)CCCCO
3360-41-6
C10H14O
150.22
MFCD00002971
LDZLXQFDGRCELX-UHFFFAOYSA-N
4-phenyl-1-butanol,benzenebutanol,4-phenylbutanol,phenylbutyl alcohol,4-phenylbutyl alcohol,1-butanol, 4-phenyl,4-phenyl butanol-1,4-phenyl butanol,unii-q5orz1321g,4-phenyl-n-butanol
76889
4-phenylbutan-1-ol
C1=CC=C(C=C1)CCCCO
Infrared Spectrum Authentic
Specific Gravity 0.975
Boiling Point 140°C (14.0mmHg)
Density 0.9750g/mL
Refractive Index 1.5200 to 1.522
Quantity 10g
Color Colorless
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C6H5(CH2)4OH
Beilstein 06, II, 487
Formula Weight 150.22
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-butanol, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-128-8

TSCA : TSCA