Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-1-buten-4-ol, 97%

Catalog No. p-7035739
Click to view available options
Quantity:
5 g
25 g

CAS: 936-58-3 | C10H12O | 148.205 g/mol

4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Phenyl-1-buten-4-ol is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 936-58-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00039617
InChI Key RGKVZBXSJFAZRE-UHFFFAOYSA-N
Synonym 1-phenyl-3-buten-1-ol, 4-phenyl-1-buten-4-ol, 1-phenyl-but-3-en-1-ol, acmc-1bdtt, upenn_abs_025, 1-phenyl-3-buten-1-ol #, benzenemethanol,a-2-propen-1-yl, benzenemethanol, .alpha.-2-propenyl
PubChem CID 220119
IUPAC Name 1-phenylbut-3-en-1-ol
SMILES C=CCC(C1=CC=CC=C1)O

Specifications

Density 0.992
Boiling Point 234°C
Flash Point 103°C (217°F)
Refractive Index 1.53
Quantity 5 g
Formula Weight 148.2
Percent Purity 97%
Chemical Name or Material 4-Phenyl-1-buten-4-ol
Hazard Category H302
Hazard Statement GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.