Learn More
4-Phenyl-2-butanone, 98%
Description
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Phenyl-2-butanone be used as an attractant for melon flies and as an odorant for soap. It is also used in the preparation of 4-oxocyclohexanecarbaldehyde derivatives.
Solubility
Not miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Density | 0.988 |
| Boiling Point | 235°C to 237°C |
| Flash Point | 105°C (221°F) |
| Refractive Index | 1.5122 |
| Quantity | 1000 g |
| Beilstein | 1907123 |
| Solubility Information | Not miscible in water. |
| Formula Weight | 148.21 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Phenyl-2-butanone |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 219-847-4 |
| RTECSNumber | EL9600000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.