Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenyl-3-butyn-2-one, 96%

Catalog No. p-7030266
Click to view available options
Quantity:
1 g
5 g

CAS: 1817-57-8 | C10H8O | 144.173 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1817-57-8
Molecular Formula C10H8O
Molecular Weight (g/mol) 144.173
MDL Number MFCD00008776
InChI Key UPEUQDJSUFHFQP-UHFFFAOYSA-N
Synonym 4-phenyl-3-butyn-2-one, 3-butyn-2-one, 4-phenyl, 1-phenyl-1-butyn-3-one, phenylbutynone, acetylphenyl-acetylene, acmc-1boyr, 4-phenyl-3-butyn-2-on, 4-phenyl-3-batyu-2-one, 4-phenyl-3-butyn-2-0ne, 4-phenyl-but-3-yn-2-one
PubChem CID 74555
ChEBI CHEBI:51731
IUPAC Name 4-phenylbut-3-yn-2-one
SMILES CC(=O)C#CC1=CC=CC=C1

Specifications

Density 0.99
Boiling Point 75°C to 76°C (0.8 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.574
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 144.17
Percent Purity 96%
Chemical Name or Material 4-Phenyl-3-butyn-2-one
Hazard Category H302
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 217-327-1
RTECSNumber ES0890000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.