Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Phenylpyridine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
939-23-1
C11H9N
155.2
MFCD00006420
JVZRCNQLWOELDU-UHFFFAOYSA-N
pyridine, 4-phenyl,p-phenylpyridine,4-phenyl-pyridine,4-azabiphenyl,unii-452kd9ycg7,4-aza-1,1'-biphenyl,phenyl-pyridine,4-phenylpyridin,4-pyridylbenzene,zlchem 480
13651
4-phenylpyridine
C1=CC=C(C=C1)C2=CC=NC=C2
939-23-1
C11H9N
155.2
MFCD00006420
JVZRCNQLWOELDU-UHFFFAOYSA-N
pyridine, 4-phenyl,p-phenylpyridine,4-phenyl-pyridine,4-azabiphenyl,unii-452kd9ycg7,4-aza-1,1'-biphenyl,phenyl-pyridine,4-phenylpyridin,4-pyridylbenzene,zlchem 480
13651
4-phenylpyridine
C1=CC=C(C=C1)C2=CC=NC=C2
Melting Point 71.0°C to 75.0°C
Packaging Glass bottle
Boiling Point 274.0°C to 275.0°C
Infrared Spectrum Authentic
Color Yellow to Beige
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (C5H4N)C6H5
Beilstein 20, 424
Solubility Information Solubility in water: insoluble in cold water. Other solubilities: soluble in ethanol and ethyl ether
Formula Weight 155.2
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Phenylpyridine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-357-4