Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Piperazinoacetophenone, 94%

Catalog No. AC159570250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 51639-48-6 | C12H16N2O | 204.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51639-48-6
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.27
MDL Number MFCD00005962
InChI Key KPXVKKBJROCIJB-UHFFFAOYSA-N
Synonym 1-4-acetylphenyl piperazine, 1-4-piperazin-1-yl phenyl ethanone, 4'-piperazinoacetophenone, 1-4-piperazin-1-ylphenyl ethanone, 4-piperazinoacetophenone, piperazin-4-ylacetophenone, 1-4-piperazin-1-yl-phenyl-ethanone, p-piperazinoacetophenone, 1-4-piperazin-1-yl-phenyl ethanone
PubChem CID 97269
IUPAC Name 1-(4-piperazin-1-ylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)N2CCNCC2

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 107°C to 110°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Quantity 25 g
Formula Weight 204.27
Percent Purity 94%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Piperazinoacetophenone, 94%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 257-332-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.