Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(piperidin-1-ylmethyl)aniline, 97%, Thermo Scientific™

Catalog No. CC45514DA
Click to view available options
:
Amber Glass Bottle; 1g

Specifications

CAS 29608-05-7
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H18N2
Quantity 1g
Synonym 4-piperidin-1-ylmethyl aniline, 4-piperidin-1-ylmethyl-phenylamine, 4-piperidin-1-ylmethyl benzenamine, 4-piperidylmethyl phenylamine, 4-1-piperidinylmethyl aniline, 4-piperidylmethyl aniline, 1-4-aminobenzyl piperidine, 4-aminobenzylpiperidine, benzenamine, 4-1-piperidinylmethyl, 4-piperidin-1-ylmethyl-aniline
InChI Key DRYFDUUAYSVNSN-UHFFFAOYSA-N
SMILES C1CCN(CC1)CC2=CC=C(C=C2)N
IUPAC Name 4-(piperidin-1-ylmethyl)aniline
Molecular Weight (g/mol) 190.29
PubChem CID 846150
Formula Weight 190.29
Percent Purity 97%
Chemical Name or Material 4-(Piperidin-1-ylmethyl)aniline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.