Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ 4 Point Chin Strap for SecureFit™ Safety Helmet
Designed with quick and smooth adjustment features for a customized fit.
$113.00 - $202.00
Specifications
Color | Black |
---|---|
For Use With (Equipment) | 3M SecureFit X5000 Series Safety Helmets |
For Use With (Application) | Construction, Heavy Infrastructure, Oil & Gas |
Material | Nylon |
Product Type | Chin Strap |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
19-071-201
|
3M™
7100207831 |
Buckle |
Case of 10 for $113.00
|
|
|||||
19-071-200
|
3M™
7100207541 |
Magnetic buckle |
Case of 10 for $202.00
|
|
|||||
Description
- Easy adjustment
- Fits 3M™ SecureFit™ X5000 Series Safety Helmets
- 3M Catalog# X5-P4PTCS1 , 3M Catalog# X5-S4PTCS1
Specifications
Black | |
Construction, Heavy Infrastructure, Oil & Gas | |
Chin Strap |
3M SecureFit X5000 Series Safety Helmets | |
Nylon |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title