Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Propylbenzoic acid, 97%, Thermo Scientific™

Catalog No. p-188195
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 2438-05-3
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.2
MDL Number MFCD00013996
InChI Key ATZHGRNFEFVDDJ-UHFFFAOYSA-M

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 140.0°C to 144.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3CH2CH2C6H4CO2H
Quantity 1 g
Synonym 4-n-propylbenzoic acid, benzoic acid, 4-propyl, 4-propyl-benzoic acid, p-n-propyl benzoic acid, p-propylbenzoic acid, unii-3p1v85e45o, 4-propylbenzoicacid, p-propyl benzoic acid, 4-propylbenzolic acid, pubchem1348
SMILES CCCC1=CC=C(C=C1)C(=O)O
IUPAC Name 4-propylbenzoic acid
PubChem CID 137601
Formula Weight 164.2
Percent Purity 97%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Propylbenzoic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
If eye irritation persists: Get medical advice/attent

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.