Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyridinemethanol 97.0+%, TCI America™
SDP

Catalog No. P0751500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0751500G 500 g
P075125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0751500G Supplier TCI America Supplier No. P0751500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 586-95-8
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006442
InChI Key PTMBWNZJOQBTBK-UHFFFAOYSA-N
Synonym 4-pyridinemethanol, 4-pyridylcarbinol, 4-hydroxymethyl pyridine, pyridine-4-methanol, 4-pyridylmethanol, 4-pyridinylmethanol, 4-pyridinecarbinol, 4-picolyl alcohol, 4-hydroxymethylpyridine, pyridin-4-yl methanol
PubChem CID 11472
IUPAC Name (pyridin-4-yl)methanol
SMILES OCC1=CC=NC=C1

Specifications

Melting Point 55°C
Boiling Point 150°C
Quantity 500 g
Formula Weight 109.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Pyridinemethanol
EINECSNumber (9)-1112
RTECSNumber UT4691000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.