Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Pyrrolidin-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Catalog No. CC55604CB
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC55604CB 250 mg
CC55604DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC55604CB Supplier Thermo Scientific Chemicals Supplier No. CC55604CB
Add to Cart

Chemical Identifiers

CAS 650628-72-1
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD07782421
InChI Key MTBYCACQSYAYCC-UHFFFAOYSA-N
PubChem CID 23090828
IUPAC Name 4-(pyrrolidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(C1)CC2=CC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-pyrrolidin-1-ylmethyl benzaldehyde, 4-1-pyrrolidinylmethyl benzaldehyde, benzaldehyde, 4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl-benzaldehyde, 1-4-formylbenzyl pyrrolidine, 4-pyrrolizinomethyl benzaldehyde, 4-pyrrolidinylmethyl benzaldehyde, 4-pyrrolidin-1-ylmethyl-benzaldehyde, benzaldehyde,4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl benzaldehyde
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzaldehyde
Hazard Statement GHS H Statement:
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.