Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Pyrrolidinopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
2456-81-7
C9H12N2
148.21
MFCD00006415
RGUKYNXWOWSRET-UHFFFAOYSA-N
4-pyrrolidinopyridine,4-pyrrolidin-1-yl pyridine,4-1-pyrrolidinyl pyridine,pyridine, 4-1-pyrrolidinyl,4-pyrrolidin-1-yl-pyridine,4-pyrrolidinylpyridine,1-4-pyridyl pyrrolidine,zlchem 148,4-pyrrolidinepyridine,4-pyrrollidinopyridine
75567
4-pyrrolidin-1-ylpyridine
C1CCN(C1)C2=CC=NC=C2
2456-81-7
C9H12N2
148.21
MFCD00006415
RGUKYNXWOWSRET-UHFFFAOYSA-N
4-pyrrolidinopyridine,4-pyrrolidin-1-yl pyridine,4-1-pyrrolidinyl pyridine,pyridine, 4-1-pyrrolidinyl,4-pyrrolidin-1-yl-pyridine,4-pyrrolidinylpyridine,1-4-pyridyl pyrrolidine,zlchem 148,4-pyrrolidinepyridine,4-pyrrollidinopyridine
75567
4-pyrrolidin-1-ylpyridine
C1CCN(C1)C2=CC=NC=C2
Boiling Point 171°C to 173°C (27.0 mbar)
Flash Point 142°C
Infrared Spectrum Authentic
Melting Point 54°C to 58°C
Color Brown to White
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Fieser 04,416; 09,400; 10,336; 13,262; 01,1049; 02,377; 03,262; 04,443; 05,597; 06,530; 07,329; 08,449; 09,420; 10,357; 11,477; 12,441; 13,21; 14,99; 15,290; 16,304; 17,189
Solubility Information Solubility in water: 3g/L (20°C). Other solubilities: soluble in acetic acid,soluble in methanol,benzene,ethyl acetate,,chloroform,acetone and dichloroethane,insoluble in hexane and cyclohexane
Formula Weight 148.21
Physical Form Crystalline Mass, Crystals and/or Chunks
Percent Purity 98%
Chemical Name or Material 4-Pyrrolidinopyridine, 98%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 219-534-2