Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-sulfamoylbenzoic acid, 98%

Catalog No. AC303220010
Click to view available options
Quantity:
1 g
5 g

CAS: 1205-30-7 | C7H6ClNO4S | 235.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1205-30-7
Molecular Formula C7H6ClNO4S
Molecular Weight (g/mol) 235.65
InChI Key FHQAWINGVCDTTG-UHFFFAOYSA-N
Synonym 4-chloro-5-sulphamoylbenzoic acid, 3-aminosulfonyl-4-chlorobenzoic acid, csba, sulfamido-3-chlorobenzoic acid, benzoic acid, 3-aminosulfonyl-4-chloro, 3-sulfamoyl-4-chlorobenzoic acid, benzoic acid, 4-chloro-3-sulfamoyl, 4-chloro-3-sulfamoyl-benzoic acid, 4-chloro-5-sulfamoylbenzoic acid, 4-chloro-3-sulphamoylbenzoic acid
PubChem CID 14568
IUPAC Name 4-chloro-3-sulfamoylbenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)S(=O)(=O)N)Cl

Specifications

Boiling Point 256.0°C to 258.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 235.65
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-sulfamoylbenzoic acid
Hazard Category GHS Signal Word: Warning
EINECSNumber 214-882-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.