Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Styryltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. S09245G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 89114-74-9
Molecular Formula C26H21N
Molecular Weight (g/mol) 347.46
MDL Number MFCD16619204
InChI Key DXYYLUGHPCHMRQ-UHFFFAOYSA-N
Synonym 4-Diphenylaminostilbene
PubChem CID 98007462
IUPAC Name N,N-diphenyl-4-(2-phenylethenyl)aniline
SMILES C(=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C
Quantity 5 g
Formula Weight 347.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Styryltriphenylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.