Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfamoylbenzoic acid, 96%, Thermo Scientific™

Catalog No. AAB2244918
Click to view available options
Quantity:
50 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Sulfamoylbenzoic acid is a moiety of sulfamyl diuretics or antihypertensive. It is used as an antiviral and an intermediate for the manufacture of frusemide which used as a diuretic and as an antihypertensive.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-41-0
Molecular Formula C7H7NO4S
Molecular Weight (g/mol) 201.196
MDL Number MFCD00007938
InChI Key UCAGLBKTLXCODC-UHFFFAOYSA-N
Synonym carzenide, 4-carboxybenzenesulfonamide, 4-aminosulfonyl benzoic acid, carzenid, dirnate, p-carboxybenzenesulfonamide, benzoic acid, 4-aminosulfonyl, 4-aminosulfonylbenzoic acid, p-sulfamylbenzoic acid, p-sulfamoylbenzoic acid
PubChem CID 8739
IUPAC Name 4-sulfamoylbenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)S(=O)(=O)N

Specifications

Melting Point ∼285°C (decomposition)
Quantity 50 g
Beilstein 1875393
Merck Index 14,1876
Formula Weight 201.2
Percent Purity 96%
Chemical Name or Material 4-Sulfamoylbenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber DH6820000
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.