Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfo-1,8-naphthalic Anhydride Potassium Salt 98.0+%, TCI America™
SDP

Catalog No. S090425G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 71501-16-1
Molecular Formula C12H5KO6S
Molecular Weight (g/mol) 316.32
MDL Number MFCD00012097
InChI Key WOHXJVJXAWXEQP-UHFFFAOYSA-M
Synonym 4-Sulfo-1,8-naphthalenedicarboxylic Anhydride Potassium Salt
PubChem CID 16211743
IUPAC Name potassium 2,4-dioxo-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(12),5(13),6,8,10-pentaene-8-sulfonate
SMILES [K+].[O-]S(=O)(=O)C1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 316.32
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfo-1,8-naphthalic Anhydride Potassium Salt
EINECSNumber (4)-1426
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.