Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfo-2,3,5,6-tetrafluorobenzoic Acid 95.0+%, TCI America™
SDP

Catalog No. s05101g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 125662-60-4
Molecular Formula C7H2F4O5S
Molecular Weight (g/mol) 274.142
InChI Key JNESNWCWZVATQY-UHFFFAOYSA-N
PubChem CID 19807654
IUPAC Name 2,3,5,6-tetrafluoro-4-sulfobenzoic acid
SMILES C1(=C(C(=C(C(=C1F)F)S(=O)(=O)O)F)F)C(=O)O

Specifications

Quantity 1 g
UN Number 1759
Formula Weight 274.14
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Sulfo-2,3,5,6-tetrafluorobenzoic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.