Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Sulfonic calix[4]arene hydrate, 95%

Catalog No. p-3779576
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

C28H24O16S4·xH2O | 744.75 g/

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C28H24O16S4·xH2O
Molecular Weight (g/mol) 744.75
MDL Number MFCD16495912

Specifications

Color Beige-Gray to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol and ethanol
Formula Weight 744.75
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 4-Sulfonic calix[4]arene hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.