Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butoxyphenol 98.0+%, TCI America™
SDP

Catalog No. p-7119650 Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23901G 1 g
B23905G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B23901G Supplier TCI America Supplier No. B23901G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2460-87-9
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD02183556
InChI Key CIICLJLSRUHUBY-UHFFFAOYSA-N
Synonym 4-tert-butoxy phenol, 4-tert-butoxyphenol, p-tert-butoxyphenol, 4-t-butoxyphenol, phenol, 4-1,1-dimethylethoxy, unii-4w56a32ou8, 4-2-methylpropan-2-yl oxy phenol, phenol, p-tert-butoxy, 4-tert-butoxy-phenol, acmc-209gdf
PubChem CID 2773621
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]phenol
SMILES CC(C)(C)OC1=CC=C(C=C1)O

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.