Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butyl-3-thiosemicarbazide, 97%

Catalog No. AAL1070403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1070403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1070403 Supplier Thermo Scientific Chemicals Supplier No. L1070403

CAS: 13431-39-5 | C5H13N3S | 147.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of antiproliferative activity of N-substituted 2-amino-5-(2, 4-dihydroxyphenyl)-1, 3, 4-thiadiazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13431-39-5
Molecular Formula C5H13N3S
Molecular Weight (g/mol) 147.24
MDL Number MFCD00041304
InChI Key ZUWRCNZOBNETMU-UHFFFAOYSA-N
Synonym n-tert-butyl hydrazinecarbothioamide, 4-tert-butyl-3-thiosemicarbazide, 3-amino-1-tert-butylthiourea, n-tert-butylhydrazinecarbothioamide, 4-tert-butylthiosemicarbazide, hydrazinecarbothioamide,n-1,1-dimethylethyl, tert-butyl amino hydrazinomethane-1-thione, acmc-20a36m, n1-tert-butyl hydrazine-1-carbothioamide, 4-1,1-dimethylethyl-3-thiosemicarbazide
PubChem CID 737275
IUPAC Name 1-amino-3-tert-butylthiourea
SMILES CC(C)(C)NC(=S)NN

Specifications

Melting Point 139°C to 140°C
Quantity 1 g
Beilstein 507219
Solubility Information Insoluble in water.
Formula Weight 147.24
Percent Purity 97%
Chemical Name or Material 4-tert-Butyl-3-thiosemicarbazide
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.