Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylcalix[4]arene-tetraacetic acid tetraethyl ester, 95%, Thermo Scientific™

Catalog No. AC290285000
Click to view available options
Quantity:
500 mg
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97600-39-0
Molecular Formula C60H80O12
Molecular Weight (g/mol) 993.288
MDL Number MFCD00145373
InChI Key HZHADWCIBZZJNV-UHFFFAOYSA-N
Synonym sodium ionophore x, 4-tert-butylcalix 4 arene-tetraacetic acid tetraethyl ester, tetraethyl 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetate, p-tert-butylcalix 4 arene tetrakis ethyl acetate, sodium ionophore x, selectophore tm , function tested, 4-t-butylcalix 4 arene-tetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-tetraacetic acid tetraet, 4-tert-butylcalix 4 arenetetraacetic acid tetraethyl ester, 4-tert-butylcalix 4 arene-o,o',o,o'-tetraacetic acid tetraethyl ester
PubChem CID 2724889
SMILES CCOC(=O)COC1=C2CC3=CC(=CC(=C3OCC(=O)OCC)CC4=C(C(=CC(=C4)C(C)(C)C)CC5=C(C(=CC(=C5)C(C)(C)C)CC1=CC(=C2)C(C)(C)C)OCC(=O)OCC)OCC(=O)OCC)C(C)(C)C

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 993.28
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 4-tert-Butylcalix[4]arene-tetraacetic acid tetraethyl ester, 95%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.