Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenyl glycidyl ether, 95%

Catalog No. p-7037250
Click to view available options
Quantity:
5 g
25 g

CAS: 3101-60-8 | C13H18O2 | 206.285 g/mol

4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-tert-Butylphenyl glycidyl ether is used in paint industry and serves as organic coating in Paint, boat Paint, building coating material. It is also used as an epoxy reactive diluent.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3101-60-8
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00005136
InChI Key HHRACYLRBOUBKM-UHFFFAOYSA-N
Synonym p-tert-butylphenyl glycidyl ether, 4-tert-butylphenyl glycidyl ether, 4-t-butylphenyl glycidyl ether, 2-4-tert-butylphenoxy methyl oxirane, tert-butylphenol glycidyl ether, tert-butylphenyl glycidyl ether, t-butyl phenyl glycidyl ether, ccris 2638, oxirane, 4-1,1-dimethylethyl phenoxy methyl, 2-4-tert-butyl phenoxy methyl oxirane
PubChem CID 18360
IUPAC Name 2-[(4-tert-butylphenoxy)methyl]oxirane
SMILES CC(C)(C)C1=CC=C(C=C1)OCC2CO2

Specifications

Density 1.038
Boiling Point 165°C to 170°C (14 mmHg)
Flash Point 102°C (216°F)
Refractive Index 1.515
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 206.28
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4-tert-Butylphenyl glycidyl ether
Hazard Category H317
Hazard Statement GHS H Statement
H315-H319-H317
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P272-P280g-P302+P352-P333+P313-P363-P501c
EINECSNumber 221-453-2
RTECSNumber TX4250000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.