Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-tert-Butylphenylboronic acid, 97%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Specifications
Color | White |
Melting Point | 191.0°C to 196.0°C |
Quantity | 5g |
Packaging | Glass bottle |
Infrared Spectrum | Authentic |
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Assay Percent Range | 96% min. (HPLC) |
Linear Formula | (CH3)3CC6H4B(OH)2 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in ether,thf,dmso,dmf,methanol |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning