Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-tert-Butylphenylmagnesium bromide, 2M solution in diethyl ether, AcroSeal™

Catalog No. ac432861000
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle

Catalog No. AC432861000

Supplier: Thermo Scientific Chemicals 432861000

Only null left
Add to Cart

CAS: 63488-10-8 | C10H13BrMg | 237.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63488-10-8, 60-29-7
Molecular Formula C10H13BrMg
Molecular Weight (g/mol) 237.42
MDL Number MFCD00013244
InChI Key YECFWFOEQLAVNH-UHFFFAOYSA-M
Synonym 4-tert-butylphenylmagnesium bromide, 4-t-butylphenylmagnesium bromide, bromo 4-tert-butylphenyl magnesium, magnesium, bromo 4-1,1-dimethylethyl phenyl, 4-tert-butylphenylmagnesium bromide solution, yecfwfoeqlavnh-uhfffaoysa-m, 4-tert-butylphenyhnagnesium bromide, 4-tert.-butylphenylmagnesium bromide, 4-tert-butylphenyl magnesium bromide, 4-tert butyl phenyl magnesium bromide, grignard reagent
PubChem CID 4608371
IUPAC Name magnesium;tert-butylbenzene;bromide
SMILES CC(C)(C)C1=CC=C([Mg]Br)C=C1

Specifications

CAS Min % 42.36, 53.19
CAS Max % 46.82, 57.64
Color Brown to Yellow
Density 1.0650g/mL
Flash Point −22°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)3CC6H4MgBr
Quantity 100 mL
Specific Gravity 1.065
Formula Weight 237.42
Physical Form Solution
Chemical Name or Material 4-tert-Butylphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Harmful if swallowed.
Extremely flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.