Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenebis(triphenylphosphine)platinum(0), 98%

Catalog No. AC369360010
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

CAS: 12120-15-9 | C38H34P2Pt | 747.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12120-15-9
Molecular Formula C38H34P2Pt
Molecular Weight (g/mol) 747.72
MDL Number MFCD00012028
InChI Key RZUASTIKPBCXPU-UHFFFAOYSA-N
IUPAC Name ethene bis(triphenylphosphane) platinum
SMILES [Pt].C=C.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 127.0°C to 131.0°C
Assay Percent Range 97.5% min. (ex Pt)
Packaging Glass bottle
Linear Formula [(C6H5)3P]2Pt[H2C=CH2]
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 747.73
Percent Purity 98%
Chemical Name or Material Ethylenebis(triphenylphosphine)platinum(0)
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.