Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Thiouridine, 98+%

Catalog No. p-7041906
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60679MC 100 mg
AAJ60679MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60679MC Supplier Thermo Scientific Chemicals Supplier No. J60679MC
Only null left
Add to Cart

CAS: 13957-31-8 | C9H12N2O5S | 260.26 g/mol

4-Thiouridine is a constituent of several transfer RNA's. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Thiouridine is a constituent of several transfer RNA′s. 4-Thiouridine is a thionucleobase used as an antisense agent. Widely used for RNA analysis including RNA-RNA cross coupling and RNA labelling. 4-Thiouridine is a photoreactive (crosslinking) uridine analogue that upon phosphorylation to 4-thioUTP may be incorporated into RNA structures. 4-Thiouridine may be used to study the specificity and kinetics of uridine-cytidine kinases. 4-Thio Uridine is typically used to modify oligos slated for RNA, or RNA-protein, structural studies. A 4-thio-rU modified RNA pentamer was used to study the effect of this modification on codon-anticodon interaction when it is in the wobble position of tRNA.

Solubility
Soluble in water (20 mg/ml), and methanol (10 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13957-31-8
Molecular Formula C9H12N2O5S
Molecular Weight (g/mol) 260.26
MDL Number MFCD00006538
InChI Key ZLOIGESWDJYCTF-XVFCMESISA-N
PubChem CID 3032615
ChEBI CHEBI:20480
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O

Specifications

Color Yellow
Quantity 100 mg
Synonym 4-thiouridine, thiouridine, uridine, 4-thio, 1-beta-d-ribofuranosyl-4-thiouracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, 1-beta-d-ribofuranosyl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, s4u, thiouridine van, uridine,4-thio, 4-thio-d-uridine
Solubility Information Soluble in water (20mg/ml),and methanol (10mg/ml).
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-sulfanylidenepyrimidin-2-one
Formula Weight 260.27
Percent Purity ≥98%
Physical Form Crystalline
Chemical Name or Material 4-Thiouridine
EINECSNumber 237-735-3
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.