Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Thiouridine, ∽98, MP Biomedicals™

Catalog No. ICN15213483 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
0.1 g
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15213483 250 mg
ICN15213405 5 mg
ICN15213480 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15213483 Supplier MP Biomedicals, Inc Supplier No. 0215213483
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13957-31-8
Molecular Formula C9H12N2O5S
Molecular Weight (g/mol) 260.26
MDL Number MFCD00006538
InChI Key ZLOIGESWDJYCTF-XVFCMESISA-N
Synonym 4-thiouridine, thiouridine, uridine, 4-thio, 1-beta-d-ribofuranosyl-4-thiouracil, 1-2r,3r,4s,5r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, 1-beta-d-ribofuranosyl-4-thioxo-3,4-dihydropyrimidin-2 1h-one, s4u, thiouridine van, uridine,4-thio, 4-thio-d-uridine
PubChem CID 3032615
ChEBI CHEBI:20480
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=CC(=S)NC1=O

Specifications

Color Yellow
Assay Percent Range ∽98
Quantity 250 mg
Formula Weight 260.3
Physical Form Powder
Chemical Name or Material 4-Thiouridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.