Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(trans-4-Propylcyclohexyl)benzoic Acid 98.0+%, TCI America™
SDP

Catalog No. P243825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P243825G 25 g
P24385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P243825G Supplier TCI America Supplier No. P243825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65355-29-5
Molecular Formula C16H22O2
Molecular Weight (g/mol) 246.35
MDL Number MFCD06658177
InChI Key VACLULPMEXHBMD-UHFFFAOYSA-N
Synonym 4-trans-4-n-propylcyclohexyl benzoic acid, 4-trans-4-propylcyclohexyl benzoic acid, 4-4-propylcyclohexyl benzoic acid, trans-4-4-n-propylcyclohexyl benzoic acid, 4-propyl cyclohexyl benzoic acid, benzoic acid, 4-4-propylcyclohexyl, 4-trans-4-n-propylcyclohexyl benzoicacid, 4-1s,4r-4-propylcyclohexyl benzoic acid, acmc-20lbd4
PubChem CID 4552940
IUPAC Name 4-(4-propylcyclohexyl)benzoic acid
SMILES CCCC1CCC(CC1)C2=CC=C(C=C2)C(=O)O

Specifications

Melting Point 206°C
Color White
Quantity 25 g
Formula Weight 246.35
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(trans-4-Propylcyclohexyl)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.