Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethoxy)aniline, 99%

Catalog No. p-205259
Click to view available options
Quantity:
2.5 g
25 g
Packaging:
Glass bottle

CAS: 461-82-5 | C7H6F3NO | 177.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-82-5
Molecular Formula C7H6F3NO
Molecular Weight (g/mol) 177.12
MDL Number MFCD00041314
InChI Key XUJFOSLZQITUOI-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy aniline, 4-trifluoromethoxy-phenylamine, 4-trifluoromethoxy benzenamine, p-trifluoromethoxyaniline, 4-trifluoroethoxy aniline, benzenamine, 4-trifluoromethoxy, unii-p40l42cvf6, alpha,alpha,alpha-trifluoro-p-anisidine, p-aminotrifluoromethoxybenzene, 4-amino-alpha,alpha,alpha-trifluoroanisole
PubChem CID 600848
IUPAC Name 4-(trifluoromethoxy)aniline
SMILES C1=CC(=CC=C1N)OC(F)(F)F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3100g/mL
Boiling Point 83.0°C to 85.0°C (15.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4620 to 1.4640
Linear Formula CF3OC6H4NH2
Quantity 2.5 g
Specific Gravity 1.31
Formula Weight 177.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-(Trifluoromethoxy)aniline, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Fatal in contact with skin.
May cause damage to organs through prolonged or repeated exposure.<b
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 207-317-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.