Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethyl)phenylacetic Acid 97.0+%, TCI America™
SDP

Catalog No. T178325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T178325G 25 g
T17831G 1 g
T17835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T178325G Supplier TCI America Supplier No. T178325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32857-62-8
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD00004352
InChI Key HNORVZDAANCHAY-UHFFFAOYSA-N
Synonym 4-trifluoromethyl phenylacetic acid, 2-4-trifluoromethyl phenyl acetic acid, 4-trifluoromethylphenylacetic acid, 4-trifluoromethyl phenyl acetic acid, alpha,alpha,alpha-trifluoro-p-tolyl acetic acid, benzeneacetic acid, 4-trifluoromethyl, p-trifluoromethyl phenylacetic acid, p-trifluoromethylphenylacetic acid, alpha,alpha,alpha-trifluoro-4-tolyl acetic acid
PubChem CID 118342
IUPAC Name 2-[4-(trifluoromethyl)phenyl]acetic acid
SMILES OC(=O)CC1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 83°C
Quantity 25 g
Formula Weight 204.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(Trifluoromethyl)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.