Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Trifluoromethylthio)benzyl bromide, 97%, Thermo Scientific™

Catalog No. AAL1920503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1920503 1 g
AAL19205MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1920503 Supplier Thermo Scientific Chemicals Supplier No. L1920503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21101-63-3
Molecular Formula C8H6BrF3S
Molecular Weight (g/mol) 271.095
MDL Number MFCD00129189
InChI Key VAYGJBCOCRVROJ-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio benzyl bromide, 4-bromomethyl phenyl trifluoromethyl sulfane, 4-trifluoromethylthio benzylbromide, 1-bromomethyl-4-trifluoromethyl sulfanyl benzene, benzene, 1-bromomethyl-4-trifluoromethyl thio, 1-bromomethyl-4-trifluoromethylsulfanyl-benzene, 1-bromomethyl-4-trifluoromethylsulfanyl benzene, acmc-1coe0, 4-trifluoromethylthiobenzyl bromide, 4 trifluoromethylthio benzyl bromide
PubChem CID 2777882
IUPAC Name 1-(bromomethyl)-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1CBr)SC(F)(F)F

Specifications

Melting Point 53°C to 56°C
Boiling Point 115°C to 118°C (13 mmHg)
Odor Pungent
Quantity 1 g
UN Number UN3261
Beilstein 2209970
Formula Weight 271.1
Percent Purity 97%
Chemical Name or Material 4-(Trifluoromethylthio)benzyl bromide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.