Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Trimethylsilyl-3-butyn-1-ol, 98%
Click to view available options
Quantity:
5 g
25 g
Description
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
4-Trimethylsilyl-3-butyn-1-ol is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Boiling Point | 97°C to 98°C (24mmHg) |
Quantity | 25 g |
Molecular Formula | C10H8N2S2 |
Linear Formula | HOCH2CH2C≡CSi(CH3)3 |
UN Number | UN1993 |
Flash Point | 60°C (140°F) |
Synonym | 2,2'-dithiodipyridine, 2,2'-dipyridyl disulfide, 1,2-di pyridin-2-yl disulfane, aldrithiol-2, 2,2'-dipyridyldisulfide, aldrithiol 2, bis 2-pyridyl disulfide, 2-pyridin-2-yldisulfanyl pyridine, 2-dipyridyl disulfide, 2-pyridinyl disulfide |
InChI Key | HAXFWIACAGNFHA-UHFFFAOYSA-N |
SMILES | C1=CC=NC(=C1)SSC2=CC=CC=N2 |
IUPAC Name | 2-(pyridin-2-yldisulfanyl)pyridine |
Show More |
Safety and Handling
Hazard Category | H226-H315-H319-H335 |
Hazard Statement | GHS H Statement H226-H315-H319-H335 Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction