Learn More
4-Tritylaniline, 97%
Description
4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Tritylaniline was used in the preparation of 5-tritylisatin, 2,7-di-tert-butyl-9,9-dimethyl-4,5-bis(4-tritylanilinocarbonyl)-9H-xanthene methanol trisolvate monohydrate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 256°C to 260°C |
Quantity | 5 g |
Beilstein | 2750328 |
Solubility Information | Insoluble in water. |
Formula Weight | 335.45 |
Percent Purity | 97% |
Chemical Name or Material | 4-Tritylaniline |
Safety and Handling
EINECSNumber | 245-346-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.